Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547403
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547403 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547403 Supplier Invitrogen™ Supplier No. PA547403
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CLECSF9 and rhCLEC4F is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4D
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CLEC4D
Gene Accession No. Q8WXI8
Gene Alias CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8
Gene Symbols CLEC4D
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8 Gly52-Asn215.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338339
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.