Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC4E Antibody (16E3), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149311AF488
Description
CLEC4E Monoclonal specifically detects CLEC4E in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
CLEC4E | |
Monoclonal | |
Alexa Fluor 488 | |
Q9ULY5 | |
CLEC4E | |
Recombinant human CLEC4E (41-219aa) purified from E. coli (NP_055173). | |
Protein G purified | |
RUO | |
Primary | |
Human CLEC4E (41-219aa) | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
16000 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
Mouse | |
25 kDa | |
0.1 mL | |
Immunology | |
26253 | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction