Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLEC4E Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116924
Description
Positive Samples: Mouse spleen, Rat spleen Immunogen sequence: SQEEQEFLSY KKPKMREFFI GLSDQVVEGQ WQWVDGTPLT KSLSFWDVGE PNNIATLEDC ATMRDSSNPR QNWNDVTCFL N.
CLEC4E encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signaling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type II transmembrane protein is a downstream target of CCAAT/enhancer binding protein (C/EBP), beta (CEBPB) and may play a role in inflammation. Alternative splice variants have been described but their full-length sequence has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
CLEC4E | |
Polyclonal | |
Unconjugated | |
CLEC4E | |
C86253; CLEC4E; CLECSF9; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 9; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 9; C-type lectin domain family 4 member E; C-type lectin domain family 4, member E; C-type lectin superfamily member 9; C-type lectin, superfamily member 9; immunoreceptor; macrophage-inducible C-type lectin; Mincle; UNQ218/PRO244 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
450223, 56619 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.79 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q67EQ1, Q9R0Q8 | |
CLEC4E | |
Recombinant fusion protein containing a sequence corresponding to amino acids 120-200 of human CLEC4E (NP_0551731). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction