Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4F Monoclonal Antibody (10-A6-1)
GREENER_CHOICE

Catalog No. PIMA536101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536101 Supplier Invitrogen™ Supplier No. MA536101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4F Monoclonal Antibody for Flow, Western Blot

Receptor with an affinity for galactose and fucose. Could be involved in endocytosis. Target protein's carbohydrate binding receptor with an affinity for fucose and galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4F
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10-A6-1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Clec4f
Gene Accession No. Q8N1N0
Gene Alias Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor
Gene Symbols Clec4f
Host Species Mouse
Immunogen Recombinant protein within Human CLEC4F aa 93-426.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.