Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540111 Supplier Invitrogen™ Supplier No. PA540111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG and IMR32 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Receptor with an affinity for galactose and fucose. Could be involved in endocytosis. Target protein's carbohydrate binding receptor with an affinity for fucose and galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4F
Applications Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7
Gene Clec4f
Gene Accession No. Q8N1N0
Gene Alias Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor
Gene Symbols Clec4f
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human CLEC4F. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.