Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4F Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5139828
Encompass
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5139828 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5139828

Supplier: Thermo Scientific PA5139828

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RVDNVNSQLQ VLGDHLGNTN ADIQMVKGVL KDATTLSLQT QMLRSSLEGT NAEIQRLKED LEKADALTFQ TLNFLKSSLE NTSIELHVLS RGLENANSEI QMLNASLETA NTQ

CLEC4F Polyclonal antibody specifically detects CLEC4F in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4F
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Clec4f
Gene Accession No. Q8N1N0
Gene Alias Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLEC4F
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.