Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4M Mouse anti-Human, Clone: OTI7D12, liquid, TrueMAB™
SDP

Catalog No. 501683178
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3178 Supplier OriGene Supplier No. TA810067
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4M (C-Type Lectin Domain Family 4 Member M) is a Protein Coding gene. Diseases associated with CLEC4M include Japanese Encephalitis and Severe Acute Respiratory Syndrome. Among its related pathways are C-type lectin receptor signaling pathway and Phagosome. Gene Ontology (GO) annotations related to this gene include calcium-dependent protein binding. An important paralog of this gene is CD209.

Specifications

Antigen CLEC4M
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CLEC4M
Gene Accession No. Q9H2X3
Gene Alias CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Gene Symbols CLEC4M
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 74-329 of human CLEC4M produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10332
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.