Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC5A Monoclonal Antibody (1B5G12)
GREENER_CHOICE

Catalog No. FERMA548722
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548722 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548722 Supplier Invitrogen™ Supplier No. MA548722
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC5A Monoclonal Antibody for Western Blot, Flow, ELISA

Functions as a positive regulator of osteoclastogenesis. Cell surface receptor that signals via TYROBP. Regulates inflammatory responses. Acts as a key regulator of synovial injury and bone erosion during autoimmune joint inflammation. Critical macrophage receptor for dengue virus serotypes 1-4. The binding of dengue virus to CLEC5A triggers signaling through the phosphylation of TYROBP, this interaction does not result in viral entry, but stimulates proinflammatory cytokine release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC5A
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1B5G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CLEC5A
Gene Accession No. Q9NY25
Gene Alias CLEC5A; Clecsf5; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 5; C-type lectin domain family 5 member A; C-type lectin domain family 5, member a; C-type lectin superfamily member 5; Ly100; Mdl1; MDL-1; myeloid DAP12-associating lectin 1; myeloid DAP12-associating lectin-1; type II transmembrane protein MDL-1
Gene Symbols CLEC5A
Host Species Mouse
Immunogen Purified recombinant fragment of human CLEC5A (AA: extra(28-188)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.