Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (14N8D7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227087IR
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.Specifications
CLEC9a | |
Monoclonal | |
DyLight 755 | |
Q6UXN8 | |
CLEC9A | |
A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. | |
0.1 mL | |
Immunology, Stem Cells | |
283420 | |
Human, Primate | |
Purified |
Western Blot | |
14N8D7 | |
Western Blot | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen for the antibody is 100% homologous in Human, Orangutan, Chimp and Rhesus Monkey, 80% in Dog, pand and Horse, 73% in Bovine and 60% homologous in Mouse. CLEC9A is a glycoprotein visualized by western blot between ∽25-45 kD. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction