Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (1F6), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP308840AF594
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
CLEC9a | |
Monoclonal | |
Alexa Fluor 594 | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
RBL-2H3 cells expressing Mouse CLEC9a fused to an HA epitope. | |
100 μg | |
Immunology, Stem Cells | |
283420 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunofluorescence | |
1F6 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction