Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (2456A), PE/Cy5.5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB67762PECY55
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Flow CytometrySpecifications
CLEC9a | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
2456A | |
Flow Cytometry | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CLEC9a, Lys57-Ile264, Accession # EDK99924 | |
0.1 mL | |
Immunology, Stem Cells | |
283420 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction