Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (30L2), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221999AF532
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.Specifications
CLEC9a | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CLEC9A | |
Human recombinant CLEC9a | |
Immunology, Stem Cells | |
283420 | |
Store at 4°C in the dark. |
Flow Cytometry | |
30L2 | |
Flow Cytometry | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction