Learn More
CLEC9a Antibody (30L2), Alexa Fluor™ 647, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP221999AF647
Catalog No. NBP221999Y
Only null left
Add to Cart
Description
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CLEC9a
Monoclonal
Alexa Fluor 647
Q6UXN8
CLEC9A
Human recombinant CLEC9a
0.1 mL
Immunology, Stem Cells
283420
Store at 4C in the dark.
IgG1
Flow Cytometry
30L2
Flow Cytometry
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only