Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (30L2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221999PE
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.Specifications
CLEC9a | |
Monoclonal | |
PE | |
Q6UXN8 | |
CLEC9A | |
Human recombinant CLEC9a | |
0.1 mL | |
Immunology, Stem Cells | |
283420 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
30L2 | |
Flow Cytometry | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction