Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (683409), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6049M
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Human samples. It is validated for Flow CytometrySpecifications
CLEC9a | |
Monoclonal | |
DyLight 594 | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse myeloma cell line NS0-derived recombinant human CLEC9a, Lys57-Val241, Accession # Q6UXN8 | |
0.5 mL | |
Immunology, Stem Cells | |
283420 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
683409 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction