Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (30L2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019282 0.025 mg
NBP221999 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019282 Supplier Novus Biologicals Supplier No. NBP2219990.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CLEC9a
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 30L2
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:250, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q6UXN8
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Gene Symbols CLEC9A
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant CLEC9a. Mouse anti human CLEC 9a Q6UXN8.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283420
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.