Learn More
Description
Specifications
Specifications
| Antigen | CLEC9a |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 8F9 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
