Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC9A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547872
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547872 Supplier Invitrogen™ Supplier No. PA547872
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CLEC9a, rhCLEC2, and rhCLEC12B is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

C-type lectin domain family 9 member A (CLEC9A) is a gene located on chromosome 12p13.2, encoding a protein that belongs to the C-type lectin receptor family. CLEC9A is primarily expressed on dendritic cells and certain subsets of macrophages, playing a pivotal role in the immune system by recognizing and binding dead cell debris, including necrotic cells. This receptor facilitates the presentation of antigens derived from dead cells to T cells, thus contributing to the initiation of immune responses. CLEC9A is involved in cross-presentation, a process crucial for adaptive immunity, particularly in the context of viral infections and cancer. Its expression and activity are of particular interest for vaccine development and immunotherapy, given its ability to enhance antigen presentation and stimulate robust immune responses. Research into CLEC9A continues to explore its therapeutic potential for modulating immune activity in various diseases, including cancer and infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC9A
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CLEC9A
Gene Accession No. Q8BRU4
Gene Alias 9830005G06Rik; CD370; Clec9a; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; Dendritic cell natural killer lectin group receptor 1; DNGR1; DNGR-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046
Gene Symbols CLEC9A
Host Species Sheep
Immunogen CHO-derived recombinant mouse CLEC9a Lys57-Ile264.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232414
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.