Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLECL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139865 Supplier Invitrogen™ Supplier No. PA5139865
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ADIKTVRTSP LELAFPLQRS VSFNFSTVHK SCPAKDWKVH KGKCYWIAET KKSWNKSQND CAINNSYLMV IQDITAMVRF Highest antigen sequence indentity to the following orthologs: Rat - 26%, Mouse - 26%.

DCAL1 is a type II transmembrane, C-type lectin-like protein expressed on dendritic cells (DCs) and B cells. It interacts with subsets of T cells as a costimulatory molecule that enhances interleukin-4 (IL4; MIM 147780) production. DCAL1 is a type II transmembrane, C-type lectin-like protein expressed on dendritic cells (DCs) and B cells. It interacts with subsets of T cells as a costimulatory molecule that enhances interleukin-4 (IL4; MIM 147780) production.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLECL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLECL1
Gene Accession No. Q8IZS7
Gene Alias CLECL1; C-type lectin like 1; C-type lectin-like 1; C-type lectin-like domain family 1; DCAL1; DCAL-1; DC-associated lectin-1; dendritic cell associated lectin 1; dendritic cell-associated lectin 1; dendritic cell-associated lectin-1; type II transmembrane protein DCAL1
Gene Symbols CLECL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLECL1. Recombinant protein control fragment (Product #RP-109053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160365
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.