Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLECL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543996 Supplier Invitrogen™ Supplier No. PA543996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FSLFLICAMA GDVVYADIKT VRTSPLELAF PLQRSVSFNF STVHKSCPAK Sequence homology: Human: 100%.

DCAL1 is a type II transmembrane, C-type lectin-like protein expressed on dendritic cells (DCs) and B cells. It interacts with subsets of T cells as a costimulatory molecule that enhances interleukin-4 (IL4; MIM 147780) production. DCAL1 is a type II transmembrane, C-type lectin-like protein expressed on dendritic cells (DCs) and B cells. It interacts with subsets of T cells as a costimulatory molecule that enhances interleukin-4 (IL4; MIM 147780) production.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLECL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLECL1
Gene Accession No. Q8IZS7
Gene Alias CLECL1; C-type lectin like 1; C-type lectin-like 1; C-type lectin-like domain family 1; DCAL1; DCAL-1; DC-associated lectin-1; dendritic cell associated lectin 1; dendritic cell-associated lectin 1; dendritic cell-associated lectin-1; type II transmembrane protein DCAL1
Gene Symbols CLECL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CLECL1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160365
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.