Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Clemastine fumarate
GREENER_CHOICESDP

Catalog No. 1453100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-531-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-531-00 Supplier Tocris Bioscience™ Supplier No. 1453/100
Add to Cart
Add to Cart
This item is not returnable. View return policy

H1 antagonist

H1-receptor antagonist. Enhances differentiation of oligodendrocyte progenitor cells into myelin basic protein-positive oligodendrocytes in vitro and promotes remyelination in vivo. Also displays antimuscarinic properties. Clinically-used antihistamine.

Chemical Identifiers

CAS 14976-57-9
Molecular Formula C25H30ClNO5

Specifications

Quantity 100 mg
Formula Weight 459.97
Purity >98%
Molecular Weight (g/mol) 459.967
Chemical Name or Material Clemastine fumarate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.