Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clenbuterol-d9 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant

Catalog No. C0811ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0811ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0811ML Supplier MilliporeSigma Supplier No. C0811ML
Only null left
Add to Cart
Add to Cart

Clenbuterol is a pharmaceutical decongestant and bronchodilator prescribed to treat asthma with illicit uses as a performance-enhancing drug.

Clenbuterol is a pharmaceutical decongestant and bronchodilator prescribed to treat asthma with illicit uses as a performance-enhancing drug. This stable-labeled internal standard is suitable for clenbutreol testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, sports testing, or forensic analysis.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clenbuterol-D9 Hydrochloride
Packaging Ampule of 1 mL
CAS 184006-60-8
Molecular Formula C12D9H9Cl2N2O HCl
Concentration 100 μg/mL in Methanol (as free base)
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Pharmaceutical
Molecular Weight (g/mol) 322.71
Quantity 1 mL
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.