Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clenbuterol Monoclonal Antibody (11F6)
GREENER_CHOICE

Catalog No. PIMA549628
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549628 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549628 Supplier Invitrogen™ Supplier No. MA549628
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Clenbuterol belongs to the group of agonists. In livestock production clenbuterol improves the meat/fat ratio in fattened animals or accelerate the growth. Up to now agonists have not been authorized as adjuvants for fattening. In addition to its lipolytic and anabolic effect, clenbuterol has a relaxing effect on non-striated musculature on which is based its therapeutic use as an antiasthmatic and a tocolytic agent. When employed as a fattening adjuvant, as compared with the therapeutic use, clenbuterol is administered in a 5 to 10 times higher dose. Therefore, it is possible that clenbuterol residues may lead to a risk for consumers after illegal administration.Using the clenbuterol monocalantibody, it is possible to detect clenbuterol and other agonists in urine, muscle and liver both rapidly and with accuracy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clenbuterol
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 11F6
Concentration 3.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of CL and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.