Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clenbuterol Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986077
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6077 Supplier Bioss Supplier No. BS4532R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clenbuterol belongs to the group of agonists. In livestock production clenbuterol improves the meat/fat ratio in fattened animals or accelerate the growth. Up to now agonists have not been authorized as adjuvants for fattening. In addition to its lipolytic and anabolic effect, clenbuterol has a relaxing effect on non-striated musculature on which is based its therapeutic use as an antiasthmatic and a tocolytic agent. When employed as a fattening adjuvant, as compared with the therapeutic use, clenbuterol is administered in a 5 to 10 times higher dose. Therefore, it is possible that clenbuterol residues may lead to a risk for consumers after illegal administration.Using the clenbuterol monocalantibody, it is possible to detect clenbuterol and other agonists in urine, muscle and liver both rapidly and with accuracy.

Specifications

Antigen Clenbuterol
Applications ELISA
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen Clenbuterol conjugated to KLH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.