Learn More
Invitrogen™ Clenoliximab Chimeric Recombinant Rabbit Monoclonal Antibody (CE9.1 (Clenoliximab))

Description
Specificity: The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 μg/mL.
Specifications
Specifications
| Antigen | Clenoliximab Chimeric |
| Applications | ELISA, Flow Cytometry, Inhibition Assays, Immunohistochemistry, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Clone | CE9.1 (Clenoliximab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene Alias | 4PE; IDEC-151 |
| Host Species | Rabbit |
| Immunogen | Domain 1 of human CD4. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.