Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clevudine, Thermo Scientific™

Catalog No. AC469120010
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 163252-36-6
Molecular Formula C10H13FN2O5
Molecular Weight (g/mol) 260.22
MDL Number MFCD00935785
InChI Key GBBJCSTXCAQSSJ-XQXXSGGOSA-N
IUPAC Name 1-[(2S,3R,4S,5S)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN([C@H]2O[C@@H](CO)[C@H](O)[C@H]2F)C(=O)NC1=O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Light Yellow
Quantity 1 g
Chemical Name or Material Clevudine
Hazard Category Exclamation mark
Recommended Storage Normal conditions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.