Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLGN Antibody (SR1007), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CLGN |
---|---|
Clone | SR1007 |
Dilution | Western Blot, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:20, Serum 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
CLGN Monoclonal antibody specifically detects CLGN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, SerumSpecifications
CLGN | |
Western Blot, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:20, Serum 1:50-1:100 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1047 | |
IgG | |
Affinity purified |
SR1007 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
calmegin | |
A synthesized peptide derived from human CLGN (Uniprot #: O14967) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title