Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLGN Monoclonal Antibody (9C8G10)
GREENER_CHOICE

Catalog No. PIMA531704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531704 Supplier Invitrogen™ Supplier No. MA531704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31704 has been tested in indirect ELISA.

Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLGN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9C8G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CLGN
Gene Accession No. O14967
Gene Alias 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P
Gene Symbols CLGN
Host Species Mouse
Immunogen Purified recombinant fragment of human CLGN (AA: 249-405) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1047
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.