Learn More
Invitrogen™ CLGN Recombinant Rabbit Monoclonal Antibody (8R8M1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537938
Description
Positive Samples: PC-3, Mouse testis, Rat testis Immunogen sequence: FGPDKCGEDY KLHFIFRHKH PKTGVFEEKH AKPPDVDLKK FFTDRKTHLY TLVMNPDDTF EVLVDQTVVN KGSLLEDVVP PIKPPKEIED PNDKKPEEWD ERAKIPDPSA VKPEDWDESE PAQIEDSSVV KPAGWLDDEP KFIPDPNAEK PDDWNEDTDG EWEAPQILNP A.
Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.
Specifications
CLGN | |
Recombinant Monoclonal | |
0.28 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14967, P52194 | |
CLGN | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-350 of human Calmegin (O14967). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
8R8M1 | |
Unconjugated | |
CLGN | |
4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P | |
Rabbit | |
Affinity chromatography | |
RUO | |
1047, 12745, 685504 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.