Learn More
Invitrogen™ CLGN Recombinant Rabbit Monoclonal Antibody (6U8D7)

Description
Positive Samples: PC-3, Mouse testis, Rat testis Immunogen sequence: FGPDKCGEDY KLHFIFRHKH PKTGVFEEKH AKPPDVDLKK FFTDRKTHLY TLVMNPDDTF EVLVDQTVVN KGSLLEDVVP PIKPPKEIED PNDKKPEEWD ERAKIPDPSA VKPEDWDESE PAQIEDSSVV KPAGWLDDEP KFIPDPNAEK PDDWNEDTDG EWEAPQILNP A.
Specifications
Specifications
| Antigen | CLGN |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 6U8D7 |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CLGN |
| Gene Accession No. | O14967, P52194 |
| Gene Alias | 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.