Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLGN Recombinant Rabbit Monoclonal Antibody (JE58-25)
GREENER_CHOICE

Catalog No. PIMA542516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542516 Supplier Invitrogen™ Supplier No. MA542516
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Jurkat cell lysate, 293T cell lysate, rat testis tissue, human fallopian tube tissue, mouse testis tissue, SH-SY5Y. Subcellular Location: Endoplasmic reticulum membrane.

Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLGN
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE58-25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLGN
Gene Accession No. O14967, P52194
Gene Alias 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P
Gene Symbols CLGN
Host Species Rabbit
Immunogen Recombinant protein within human Calmegin aa 200-350 (Lumenal).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1047, 12745, 685504
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.