Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLHC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560994
Description
Immunogen sequence: LTKYMKHLED KYAEIKQAML IKYVPAQRKA DLDEEMIVLL KRRDVAENLN KKLQFCHQRL QIISQALSSW VKSDMSSPFQ DFVEQIQKTK Y Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.
CLHC1 is a protein coding gene.
Specifications
CLHC1 | |
Polyclonal | |
Unconjugated | |
CLHC1 | |
C2orf63; clathrin heavy chain linker domain containing 1; clathrin heavy chain linker domain-containing protein 1; CLHC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
130162 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NHS4 | |
CLHC1 | |
Recombinant protein corresponding to Human CLHC1. Recombinant protein control fragment (Product #RP-99689). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction