Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119713 Supplier Invitrogen™ Supplier No. PA5119713
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse kidney tissue lysates, A431, MG-63, SKOV-3, rat large intestine tissue, human kidney tissue, human placenta tissue, mouse colon tissue. Subcellular Location: Cell membrane, Cytoplasm, Membrane, Nucleus.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family; the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CLIC1
Gene Accession No. O00299, Q6MG61, Q9Z1Q5
Gene Alias Chloride channel ABP; chloride intracellular channel 1; chloride intracellular channel protein; chloride intracellular channel protein 1; chloride intracellular channel protein 1 {ECO:0000250; Clcp; CLIC1; clic1 {ECO:0000312; CLIC1 protein; CLIC1; NCC27; DADB-110M10.1; EMBL:CAE83985.1, ECO:0000312; EMBL:CAE83985.1}; FLJ94671; G6; hRNCC; hypothetical protein LOC324481; NCC27; Nuclear chloride ion channel 27; nuclear chloride ion channel protein; p64CLCP; regulatory nuclear chloride ion channel protein; RGD:1303043}; RNCC protein; UniProtKB:O00299, ECO:0000312; wu:fc30e04; zgc:77044
Gene Symbols CLIC1
Host Species Rabbit
Immunogen Recombinant protein within Human CLIC1 aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114584, 1192, 406864
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.