Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537851
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537851 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537851 Supplier Invitrogen™ Supplier No. PA537851
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, rat, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family; the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CLIC1
Gene Accession No. O00299
Gene Alias Chloride channel ABP; chloride intracellular channel 1; chloride intracellular channel protein; chloride intracellular channel protein 1; chloride intracellular channel protein 1 {ECO:0000250; Clcp; CLIC1; clic1 {ECO:0000312; CLIC1 protein; CLIC1; NCC27; DADB-110M10.1; EMBL:CAE83985.1, ECO:0000312; EMBL:CAE83985.1}; FLJ94671; G6; hRNCC; hypothetical protein LOC324481; NCC27; Nuclear chloride ion channel 27; nuclear chloride ion channel protein; p64CLCP; regulatory nuclear chloride ion channel protein; RGD:1303043}; RNCC protein; UniProtKB:O00299, ECO:0000312; wu:fc30e04; zgc:77044
Gene Symbols CLIC1
Host Species Goat
Immunogen Peptide with sequence C-TVDTKRRTETVQK, from the internal region of CLIC1 (aa45-57).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1192
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.