Learn More
Invitrogen™ CLIC1 Polyclonal Antibody

Description
Immunogen sequence: NVTTVDTKRR TETVQKLCPG GQLPFLLYGT EVHTDTNKIE EFLEAVLCPP RYPKLAALNP ESNTAGLDIF AKFSAYIKNS NPALNDNLEK GLLKALKVLD NYLTSPLPEE VDETSAEDEG VSQRKFLDGN ELTLADCNLL PKLHIVQVVC KKYRGFTIPE AFRGVHRYLS NAYAREEFAS TCPDDEEIEL AYEQVAKALK.
Specifications
Specifications
| Antigen | CLIC1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.58 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CLIC1 |
| Gene Accession No. | O00299, Q6MG61, Q9Z1Q5 |
| Gene Alias | Chloride channel ABP; chloride intracellular channel 1; chloride intracellular channel protein; chloride intracellular channel protein 1; chloride intracellular channel protein 1 {ECO:0000250; Clcp; CLIC1; clic1 {ECO:0000312; CLIC1 protein; CLIC1; NCC27; DADB-110M10.1; EMBL:CAE83985.1, ECO:0000312; EMBL:CAE83985.1}; FLJ94671; G6; hRNCC; hypothetical protein LOC324481; NCC27; Nuclear chloride ion channel 27; nuclear chloride ion channel protein; p64CLCP; regulatory nuclear chloride ion channel protein; RGD:1303043}; RNCC protein; UniProtKB:O00299, ECO:0000312; wu:fc30e04; zgc:77044 |
| Gene Symbols | CLIC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.