Learn More
Invitrogen™ CLIC2 Monoclonal Antibody (A5G9)

Description
Positive Control: K562 cell lysates, human liver tissue lysate, human skeletal muscle tissue lysate, HepG2, human fetal skeletal muscle tissue, human liver tissue, SH-SY5Y. Subcellular Location: Cytoplasm, Membrane.
Specifications
Specifications
| Antigen | CLIC2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A5G9 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CLIC2 |
| Gene Accession No. | O15247 |
| Gene Alias | chloride intracellular channel 2; chloride intracellular channel protein 2; Chloride intracellular channel protein 2-like protein; CLIC2; CLIC2b; MRXS32; XAP121 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.