Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541005 Supplier Invitrogen™ Supplier No. PA541005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SAEEPPVSRR LFLDGDQLTL ADCSLLPKLN IIKVAAKKYR DFDIPAEFSG Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 100%.

Chloride intracellular channel 2 is a member of the p64 family, a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. CLIon Channel 2 encodes a protein that is detected in fetal liver and adult skeletal muscle tissue. CLIon Channel2 maps to the candidate region on chromosome X for incontinentia pigmenti.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLIC2
Gene Accession No. O15247
Gene Alias chloride intracellular channel 2; chloride intracellular channel protein 2; Chloride intracellular channel protein 2-like protein; CLIC2; CLIC2b; MRXS32; XAP121
Gene Symbols CLIC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CLIC2 (aa 162-211).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1193
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.