Learn More
Invitrogen™ CLIC3 Monoclonal Antibody (2C5)

Description
Peptide Sequence: RLDSYLRAPL EHELAGEPQL RESRRRFLDG DRLTLADCSL LPKLHIVDTV CAHFRQAPIP AELRGVRRYL DSAMQEKEFK YTCPHSAEIL AAYRPAVHPR.
Specifications
Specifications
| Antigen | CLIC3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2C5 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CLIC3 |
| Gene Accession No. | O95833 |
| Gene Alias | 2300003G24Rik; chloride intracellular channel 3; chloride intracellular channel protein 3; CLIC3; RP11-229P13.3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.