Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590398 Supplier Invitrogen™ Supplier No. PA590398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPPDFPSLAP RYRESNTAGN DVFHKFSAFI KNPVPAQDEA LYQQLLRALA RLDSYLRAPL EHELAGEPQL RESRRRFLDG DRLTLADCSL LPKLHIVDTV CAHFRQAPIP AELRGVRRYL DSAMQEKEFK YTCPHSAEIL AAYRPAVHPR; Positive Samples: HeLa, Mouse lung, Rat lung; Cellular Location: Cytoplasm, Membrane, Nucleus, Single-pass membrane protein.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 3 is a member of the p64 family and is predominantly localized in the nucleus and stimulates chloride ion channel activity. In addition, this protein may participate in cellular growth control, based on its association with ERK7, a member of the MAP kinase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLIC3
Gene Accession No. O95833, Q9D7P7
Gene Alias 2300003G24Rik; chloride intracellular channel 3; chloride intracellular channel protein 3; CLIC3; RP11-229P13.3
Gene Symbols CLIC3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 87-236 of human CLIC3 (NP_0046602).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296566, 69454, 9022
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.