Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC4 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299969F
Description
CLIC4 Polyclonal specifically detects CLIC4 in Human samples. It is validated for ELISA.Specifications
CLIC4 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
25932 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
chloride intracellular channel 4, chloride intracellular channel 4 like, chloride intracellular channel protein 4, CLIC4L, DKFZp566G223, FLJ38640, H1, huH1, Intracellular chloride ion channel protein p64H1, MTCLIC, p64H1 | |
Produced in rabbits immunized with purified, recombinant CLIC4 (Q9Y696-1; Ala 2-Lys 253). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction