Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC4 Mouse anti-Human, Clone: 17CT9.2.8, Abnova™

Catalog No. 89933975 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-975 Supplier Abnova Corporation Supplier No. MAB12277
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human CLIC4.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells). [provided by RefSeq]

Specifications

Antigen CLIC4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 17CT9.2.8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CLIC4.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene Alias CLIC4L/DKFZp566G223/FLJ38640/H1/MTCLIC/huH1/p64H1
Gene Symbols CLIC4
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human CLIC4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25932
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.