Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584469 Supplier Invitrogen™ Supplier No. PA584469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MALSMPLNGL KEEDKEPLIE LFVK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLIC4
Gene Accession No. Q9Y696
Gene Alias chloride intracellular channel 4; chloride intracellular channel 4 (mitochondrial); chloride intracellular channel 4 like; chloride intracellular channel protein 4; Clic4; CLIC4L; D0Jmb3; DKFZP566G223; FLJ38640; H1; huH1; intracellular chloride ion channel protein p64H1; mc3s5; mc3s5/mtCLIC; mitochondrial chloride intracellular channel 4; MTCLIC; p64H1; TU-74
Gene Symbols CLIC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLIC4. Recombinant protein control fragment (Product #RP-103762).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.