Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC4 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. ABS2088MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2088MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2088MI Supplier MilliporeSigma Supplier No. ABS2088

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Detect Chloride intracellular channel protein 4 using this rabbit polyclonal Anti-CLIC4 used in Western Blotting and Immunofluorescence, Western Blotting, Immunohistochemistry (Paraffin) and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC4
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CLIC4
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 158 amino acids from the internal region of bovine Chloride intracellular channel protein 4 (CLIC4).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001073687
Test Specificity This polyclonal antibody detects intracellular channel protein 4 in rat brain.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.