Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC5 Antibody (CLIC5-02), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262197JF549
Description
CLIC5 Monoclonal specifically detects CLIC5 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CLIC5 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLIC5 | |
KLH-conjugated peptide corresponding to the amino acids 160-173 of human CLIC5a | |
0.1 mL | |
53405 | |
Human | |
IgG2b κ |
Western Blot, Flow Cytometry | |
CLIC5-02 | |
Western Blot, Flow Cytometry | |
chloride intracellular channel 5, chloride intracellular channel 5A, chloride intracellular channel protein 5, CLIC5B, dJ447E21.4, FLJ90663, MST130, MSTP130 | |
Mouse | |
Protein A purified | |
Primary | |
The antibody TSH-51 reacts with human thyroid stimulating hormone (hTSH, thyrotropin), a glycoprotein hormone produced by the anterior pituitary gland cells in response to signals from the hypothalamus gland in the brain. Following cross-reactivity expressed as binding of labelled hormone (% of total) was determined by solid phase RIA with excess of the antibody TSH-51: hTSH (68.6), hCG (0.03), hLH (2.99), hFSH (0.66). | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction