Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119978 Supplier Invitrogen™ Supplier No. PA5119978
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Human lung tissue lysates, mouse small intestine tissue. Subcellular Location: Cytoplasm, cytoskeleton, apical cell membrane, cell cortex, membrane; centrosome, golgi apparatus.

Clic5 encodes a protein that is required for normal hearing and is necessary for the formation of stereocilia in the inner ear and normal development of the organ of Corti. Clic5 can insert into membranes and form poorly selective ion channels that may also transport chloride ions. Clic5 may play a role in the regulation of transepithelial ion absorption and secretion, and is required for the development and/or maintenance of the proper glomerular endothelial cell and podocyte architecture. The Clic5 gene encodes a member of the chloride intracellular channel (CLIC) family of chloride ion channels. The protein encoded by Clic5 associates with actin-based cytoskeletal structures and may also play a role in multiple processes including hair cell stereocilia formation, myoblast proliferation and glomerular podocyte and endothelial cell maintenance. Alternatively spliced transcript variants encoding multiple isoforms have been observed for the Clic5 gene. Diseases associated with CLIC5 include Deafness, Autosomal Recessive 103 and Autosomal Recessive Non-Syndromic Sensorineural Deafness Type Dfnb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CLIC5
Gene Accession No. Q8BXK9, Q9NZA1
Gene Alias 5730531E12Rik; B330005L24; chloride channel protein; chloride channel protein p64; chloride intracellular channel 5; chloride intracellular channel protein 5; Chlorine channel protein p64; CLIC5; DFNB102; DFNB103; Gm322; jbg; MST130; MSTP130
Gene Symbols CLIC5
Host Species Rabbit
Immunogen Recombinant protein within human CLIC5 aa 41-240.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224796, 53405
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.