Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-IT™ L-Homopropargylglycine (HPG)
GREENER_CHOICE

Catalog No. c10186
Encompass_Preferred
Click to view available options
Quantity:
5 mg

Less hazardous technique for the detection of nascent protein synthesis

Click-iT homopropargylglycine (HPG) provides a fast, sensitive, non-toxic and most importantly non-radioactive alternative to the traditional technique, 35S methionine, for the detection of nascent protein synthesis. HPG is an amino acid analog that contains a very small modification, specifically an alkyne moiety that can be fed to cultured cells and incorporated into proteins during active protein synthesis. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the alkyne modified protein is detected with an azide-containing dye or hapten together with either the Click-iT Cell Reaction Buffer Kit or the Click-iT Protein Buffer Kit. With the Click-iT Cell Reaction Buffer Kit, cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis. With the Click-iT Protein Reaction Buffer Kit, achieve detection sensitivity in 1-D gels and western blots in the low femtomole range or perform LC-MS/MS and MALDI MS analysis.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Green Features Less hazardous
Product Type L-Homopropargylglycine
Quantity 5 mg
Content And Storage Store at ≤-20°C and desiccated.
Labeling Method Metabolic Labeling
Shipping Condition Room Temperature
Product Line Click-iT™, Molecular Probes™
Label or Dye Alkyne
Format Solid
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.