Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Clindamycin hydrochloride, 97%

Catalog No. ac443510100
Click to view available options
Quantity:
10 mg

Clindamycin hydrochloride, CAS # 21462-39-5, is a lincosamide antibiotic. | CAS: 21462-39-5 | C18H34Cl2N2O5S | 461.439 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Clindamycin is a lincosamide antibiotic derived from lincomycin that has antibiotic activity against aerobic gram-positive cocci, and parasitic protozoa
  • Clindamycin reversibly binds to the 50s bacterial ribosomal subunit, thus inhibiting protein synthesis

Applications

  • Clindamycin has been used to enhance bacterial colonization in research mice during the study of commensal gut microbiota
  • Clindamycin has been used to study biofilm-mediated antibiotic resistance in an in vitro model of G. vaginalis
  • Clindamycin was used as a selection agent to isolate, enumerate, and identify Lactobacillus spp. from probiotic dairy products
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21462-39-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
PubChem CID 134129519
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin Hydrochloride
Melting Point 156.0°C to 159.0°C
Infrared Spectrum Authentic
Water 6% max.
Quantity 10 mg
Solubility Information Solubility in water: ca 50g/L. Other solubilities: soluble in pyridine, ethanol, dmf
Formula Weight 461.45
Percent Purity 97%
Chemical Name or Material Clindamycin hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.