Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin, White Powder, MP Biomedicals™

Catalog No. icn19116810
Click to view available options
Quantity:
10 mg
50 mg
100 mg

Chemical Identifiers

CAS 21462-39-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
Synonym (7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
PubChem CID 134129519
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin
Physical Form Solid
Color White
Quantity 10 mg
Formula Weight 461.4
Chemical Name or Material Clindamycin
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.