Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Clinical Chemistry Standards

Catalog No. p-4527671
Click to view available options
Quantity:
6 x 5 mL
1 x 10 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Perform in vitro diagnosis with ammonia, HDL cholesterol and lithium clinical chemistry standards.

Perform in vitro diagnosis with Thermo Scientific™ Clinical Chemistry Standards. Product include standards for ammonia, HDL cholesterol and lithium. All standards are manufactured to exacting tolerances and, wherever possible, calibrated against National Institute of Standards and Technology (NIST™) materials.

  • Most standards bottled with tapered dropper tips to guard against contamination and evaporation losses during handling and storage
  • For in vitro diagnostic use only
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 25°C
Description Clinical Chemistry Standard calibrated against NIST(National Institute of Standards and Technology) materials
Concentration 100, 200, 300 μg/dL
Detectable Analytes Ammonia, HDL Cholesterol, Lithium
Sample Type Plasma, Serum, Urine
Type Control, Standards
Quantity 6 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.