Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ Liquid QC™ Lipid Control

Catalog No. p-4423804 Shop All CLINIQA Products
Click to view available options
Level:
1
2
This item is not returnable. View return policy
Human source material. Treat as potentially infectious

Catalog No. 22-100-51

Supplier: Cliniqa™ 91501S

This item is not returnable. View return policy
Human source material. Treat as potentially infectious

Assayed quality control material for lipids

  • Human serum based lipid control
  • Assists in monitoring of analytical systems within clinical range
  • Assayed quality control material for Apolipoprotein A-1, Apolipoprotein B, Cholesterol (Total), High Density Lipoprotein, Low Density Lipoprotein, Triglycerides
  • Needs no reconstitution or frozen storage
  • Liquid stable, two levels

Certifications

CE marked

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Detectable Analytes Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, TG
Type Lipid Control
Intended Use Monitoring test procedures used for Lipid analysis
Level 1
Quantity 6 x 3 mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.